Learn More
Bellco Glass, Inc. Gas Blending Stand,1 In/4 Out
SDP
Catalog No. 501946335
Encompass_Preferred
Single air line in splits to 4 air lines out 4 hose barbs for introduction of air into 4 separate flasks. 4 gases(4) rotometers(1) tripod stand(4) 1/4" barb x 1/8"mnpt(3) branch t's" 1/8" fnpt x 1/4" tube fittings(1) 1/8" mnpt x 1/4" tube female fitting
Catalog No. 50-194-6335 Supplier Bellco Glass, Inc. Supplier No. 780333000
Single air line in splits to 4 air lines out 4 hose barbs for introduction of air into 4 separate flasks. 4 gases(4) rotometers(1) tripod stand(4) 1/4" barb x 1/8"mnpt(3) branch t's" 1/8" fnpt x 1/4" tube fittings(1) 1/8" mnpt x 1/4" tube female fitting