Learn More
Gas Burette Solution, Reagents
Gas Burette Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$197.00
Specifications
| Chemical Name or Material | Gas Burette Solution |
|---|---|
| Color | Pink |
| CAS | 7732-18-5 , 7757-82-6 |
| Grade | Reagent |
| Specific Gravity | 1.085 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-335-164
|
Reagents Holdings Llc
CG1049001C |
1 L |
Each of 1 for $197.00
|
|
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.