Learn More
Agilent Technologies Gas Clean SCD purifier
Catalog No. NC1713977
Encompass
Outlet Concentration: Moisture 0.1 ppm, Sulfur 0.1 ppm ; Filter Capacity: 2-7 g Sulfur compounds, depending on impurities. mL
Catalog No. NC1713977 Supplier Agilent Technologies Supplier No. CP17989
Outlet Concentration: Moisture 0.1 ppm, Sulfur 0.1 ppm ; Filter Capacity: 2-7 g Sulfur compounds, depending on impurities. mL