Learn More
MSC Dirt Killer Pressure Washer, 3500 PSI, 4.2 GPM, Gas, Cold Water
Catalog No. 502836268
Encompass
The Dirt Killer H360E is a robust gas-powered cold water pressure washer for industrial and commercial applications. Providing 3500 max adjustable PSI and 4.2 GPM, it efficiently cleans decks, vehicles, and heavy equipment, supporting multi-hour operation for intensive tasks.
  • Pump offers multi-hour bypass and run-dry capability, self-priming.
  • Industrial turbo nozzle for enhanced cleaning.
  • Adjustable pressure unloader.
  • Stainless steel frame with pneumatic tires.
  • Gear-drive engine for extended pump life.
  • Electric start.
  • Low oil alert, low-tone muffler, and brass inlet water filter.
  • 50 ft steel braided pressure hose.
  • Professional trigger gun with 36 in steel lance.
  • Adjustable chemical injector.
  • Four flat tip nozzle set.
Catalog No. 50-283-6268 Supplier MSC Supplier No. 61602850
Add to Cart
Add to Cart
Request a Quote
The Dirt Killer H360E is a robust gas-powered cold water pressure washer for industrial and commercial applications. Providing 3500 max adjustable PSI and 4.2 GPM, it efficiently cleans decks, vehicles, and heavy equipment, supporting multi-hour operation for intensive tasks.
  • Pump offers multi-hour bypass and run-dry capability, self-priming.
  • Industrial turbo nozzle for enhanced cleaning.
  • Adjustable pressure unloader.
  • Stainless steel frame with pneumatic tires.
  • Gear-drive engine for extended pump life.
  • Electric start.
  • Low oil alert, low-tone muffler, and brass inlet water filter.
  • 50 ft steel braided pressure hose.
  • Professional trigger gun with 36 in steel lance.
  • Adjustable chemical injector.
  • Four flat tip nozzle set.