Learn More
MSC Value Collection Pressure Washer, Gas, Cold Water, 4.5 GPM, 5000 PSI
Catalog No. 502836611
Encompass
This gas-powered, cold water pressure washer from Value Collection is engineered for demanding cleaning applications. It combines a powerful engine with a high-performance pump to deliver reliable and effective cleaning results.
  • Cold water operation
  • Gas-powered for heavy-duty use
  • 627.0 cc engine displacement
  • 4.5 GPM flow rate
  • Vanguard engine for consistent power
  • General Triplex Ceramic Plunger pump
  • 50 feet hose length
  • 5000 PSI maximum pressure
  • 3/8 inch hose size
  • Equipped with a 2-wheel roll cage frame
Catalog No. 50-283-6611 Supplier MSC Supplier No. 48352702
Add to Cart
Add to Cart
Request a Quote
This gas-powered, cold water pressure washer from Value Collection is engineered for demanding cleaning applications. It combines a powerful engine with a high-performance pump to deliver reliable and effective cleaning results.
  • Cold water operation
  • Gas-powered for heavy-duty use
  • 627.0 cc engine displacement
  • 4.5 GPM flow rate
  • Vanguard engine for consistent power
  • General Triplex Ceramic Plunger pump
  • 50 feet hose length
  • 5000 PSI maximum pressure
  • 3/8 inch hose size
  • Equipped with a 2-wheel roll cage frame