Learn More
MSC PRO-SOURCE Pressure Washer, 4000 PSI, 4 GPM, Cold Water, Gas Powered
Catalog No. 502837709
Encompass
This pressure washer offers powerful and efficient cleaning for industrial and commercial applications. Designed for heavy-duty use, it combines high pressure with a significant flow rate to tackle tough dirt and grime.
  • Provides a flow rate of 4 GPM
  • Operates with cold water
  • Gas-powered for portability and robust performance
  • Equipped with a durable General Triplex Ceramic Plunger pump
  • Includes a 50 ft hose for extended reach
  • Delivers 4000 PSI of pressure for effective cleaning
  • Features a 392 cc engine for reliable operation
  • Designed with a 2 wheel cart frame for easy mobility
  • Comes with a 3/8 inch hose
Catalog No. 50-283-7709 Supplier MSC Supplier No. 48352769
Add to Cart
Add to Cart
Request a Quote
This pressure washer offers powerful and efficient cleaning for industrial and commercial applications. Designed for heavy-duty use, it combines high pressure with a significant flow rate to tackle tough dirt and grime.
  • Provides a flow rate of 4 GPM
  • Operates with cold water
  • Gas-powered for portability and robust performance
  • Equipped with a durable General Triplex Ceramic Plunger pump
  • Includes a 50 ft hose for extended reach
  • Delivers 4000 PSI of pressure for effective cleaning
  • Features a 392 cc engine for reliable operation
  • Designed with a 2 wheel cart frame for easy mobility
  • Comes with a 3/8 inch hose