Learn More
MSC PRO-SOURCE Pressure Washer, 2.5 GPM, Gas, Cold Water, 2000 psi
Catalog No. 502838130
Encompass
This pressure washer operates with cold water and is powered by gas. It delivers a flow rate of 2.5 GPM and a pressure of 2000 psi. Featuring a Briggs & Stratton engine and a heavy-duty axial cam pump, it is equipped with a 50 feet hose and a 2-wheel cart frame for easy mobility.
  • Operates with cold water
  • Gas-powered
  • 2.5 GPM flow rate
  • Heavy-duty axial cam pump
  • 50 feet hose length
  • 2-wheel cart frame
  • 2000 psi pressure
  • 3/8 hose size
Catalog No. 50-283-8130 Supplier MSC Supplier No. 48352314
This pressure washer operates with cold water and is powered by gas. It delivers a flow rate of 2.5 GPM and a pressure of 2000 psi. Featuring a Briggs & Stratton engine and a heavy-duty axial cam pump, it is equipped with a 50 feet hose and a 2-wheel cart frame for easy mobility.
  • Operates with cold water
  • Gas-powered
  • 2.5 GPM flow rate
  • Heavy-duty axial cam pump
  • 50 feet hose length
  • 2-wheel cart frame
  • 2000 psi pressure
  • 3/8 hose size