Learn More
Chemglass Life Sciences 250mL Gas Sampling Tube, 4mm Stpk
SDP
Catalog No. CG180802
Encompass_Preferred
250mL Gas Sampling Tube, 4mm Stpk. Tube is designed for the collection and storage of gas samples. The 250ml tubes have a sidearm O.D. of 13mm, but with the addition of a syringe sampling port for use with a CG-3022-05 or CG-3022-22 septa. Supplied complete with one CG-3022-05 red rubber septa.
Catalog No. CG180802 Supplier Chemglass Life Sciences Supplier No. CG180802
250mL Gas Sampling Tube, 4mm Stpk. Tube is designed for the collection and storage of gas samples. The 250ml tubes have a sidearm O.D. of 13mm, but with the addition of a syringe sampling port for use with a CG-3022-05 or CG-3022-22 septa. Supplied complete with one CG-3022-05 red rubber septa.