Learn More
Grainger OHAUS Gas Cylinder Bench Clamp, 1"
Catalog No. NC2207446
Encompass_Preferred
The OHAUS Gas Cylinder Bench Clamp is a heavy-duty clamp designed to securely hold gas cylinders weighing up to 500 lbs to workbenches or tables. It is constructed from high-strength cast iron with a durable powder coat finish. The clamp has a 4 inch throat depth and opens to fit cylinders from 4 to 10 inches in diameter. It secures cylinders with a threaded handwheel that tightens the clamp to the desired grip. This provides a safe, stable hold for gas cylinders during use. The clamp's base is pre-drilled for easy mounting to workbenches up to 2.5 inches thick.
This item is not returnable. View return policy
Catalog No. NC2207446 Supplier Grainger Supplier No. 404R89
The OHAUS Gas Cylinder Bench Clamp is a heavy-duty clamp designed to securely hold gas cylinders weighing up to 500 lbs to workbenches or tables. It is constructed from high-strength cast iron with a durable powder coat finish. The clamp has a 4 inch throat depth and opens to fit cylinders from 4 to 10 inches in diameter. It secures cylinders with a threaded handwheel that tightens the clamp to the desired grip. This provides a safe, stable hold for gas cylinders during use. The clamp's base is pre-drilled for easy mounting to workbenches up to 2.5 inches thick.