Learn More
Grainger JAMCO Cylinder Storage Cabinet, Steel, 6 Vertical Cylinders, 1 ea.
Catalog No. 502786245
Encompass_Preferred
This cylinder storage cabinet is designed for the safe, vertical storage of up to six compressed gas cylinders. Featuring an all-welded steel construction with a powder-coated safety yellow finish, it provides a durable solution for organizing cylinders in industrial or laboratory environments. The cabinet is designed to meet applicable safety standards for workplace storage.
  • Holds up to 6 vertical gas cylinders
  • Compatible with tank sizes from 20 lb to 43 lb
  • All-welded construction with 13 gauge flattened mesh sides
  • Solid 14 gauge steel roof
  • Manual close doors with slide bolt latch for secure access
  • Includes safety signage in large red letters
  • Features foot pads with pre-drilled holes for mounting
  • Ready to use, fully assembled design
Catalog No. 50-278-6245 Supplier Grainger Supplier No. 18G968
This cylinder storage cabinet is designed for the safe, vertical storage of up to six compressed gas cylinders. Featuring an all-welded steel construction with a powder-coated safety yellow finish, it provides a durable solution for organizing cylinders in industrial or laboratory environments. The cabinet is designed to meet applicable safety standards for workplace storage.
  • Holds up to 6 vertical gas cylinders
  • Compatible with tank sizes from 20 lb to 43 lb
  • All-welded construction with 13 gauge flattened mesh sides
  • Solid 14 gauge steel roof
  • Manual close doors with slide bolt latch for secure access
  • Includes safety signage in large red letters
  • Features foot pads with pre-drilled holes for mounting
  • Ready to use, fully assembled design