Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Gas Cylinder Lockout

Catalog No. 19103973 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-103-973 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-103-973 Supplier Brady™ Supplier No. LM023E
Add to Cart
Add to Cart

Safely lockout gas cylinders to help avoid workplace accidents. The Brady™ Gas Cylinder Lockout is adjustable and installs in seconds. There is no need to match your lockout device to varying diameters or threads of cylinder caps.

  • No need to match your lockout device to varying diameters and threads of cylinder caps
  • Adjustable and installs in just seconds
  • Device is manufactured to accommodate rings up to 3.5 in. diameter

Specifications

Alternate Part Number ASSY, GAS CAP CYLINDER LO
Color Red
Material Polypropylene
Height (Metric) 158.80 mm
Quantity 1 Ea.

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.