Learn More
Justrite Gas-Cylinder Support Brackets
Catalog No. 19085818 Shop All Justrite Safety Group Products
Change view
Click to view available options
Holds:
1 Cylinder
4 Cylinders
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
19-085-818 1 Cylinder
19-085-824 4 Cylinders
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-085-818 Supplier Justrite Supplier No. 35260
Add to Cart
Add to Cart
Secure multiple gas-cylinders to wall or bench for safe and compliant storage.
  • Brackets accept cylinder sizes ranging from 4 to 12 in. (102 to 305 mm) and 9 to 16 in. (229 to 407 mm)
  • Bumper guard edging with steel reinforced vinyl protects cylinders from damage while providing extra grip
  • Constructed from heavy-gauge powder-coated steel, or unfinished stainless steel
  • For bench mounting, use models to clamp to tables from 1 to 2.75 in. (26 to 70 mm) thick
  • Heavy-duty polypropylene support straps, 1.5 in. (38 mm) wide and rated at 1138 lb. (516 kg), feature steel cinch buckle and hold individual cylinders securely against bracket
  • Stainless steel brackets use a nylon side latch buckle to protect against corrosion
Specifications
Type Gas Cylinder Stand
Holds 1 Cylinder
Load Bearing Capacity (English) 1000 lb.
Load Bearing Capacity (Metric) 454 kg
Material Stainless Steel