Learn More
Grainger Gas Cylinder Truck, 42in
Catalog No. NC1466390
Encompass_Preferred
The Grainger Gas Cylinder Truck is a heavy-duty welded steel construction cylinder truck designed for safe and easy transport of high-pressure gas cylinders. It features a 42 inch high backrack to cradle cylinders, a powder coat finish to resist corrosion, 8 inch rubber wheels, and a 500 lb weight capacity. The 42 x 24 inch platform accommodates most cylinder sizes. This industrial-grade cylinder truck provides a stable, mobile solution for gas cylinder handling and transportation needs.
Catalog No. NC1466390 Supplier Grainger Supplier No. 34D674
The Grainger Gas Cylinder Truck is a heavy-duty welded steel construction cylinder truck designed for safe and easy transport of high-pressure gas cylinders. It features a 42 inch high backrack to cradle cylinders, a powder coat finish to resist corrosion, 8 inch rubber wheels, and a 500 lb weight capacity. The 42 x 24 inch platform accommodates most cylinder sizes. This industrial-grade cylinder truck provides a stable, mobile solution for gas cylinder handling and transportation needs.