Learn More
Chemglass Life Sciences Gas Dispersion Adapter, Fritted, for 20 L Gas Scrubber
SDP
Catalog No. 502733575
Encompass_Preferred
Fritted gas dispersion adapter designed to introduce and evenly disperse incoming gas into the liquid phase of a 20 L gas scrubber, improving gas-liquid contact for scrubbing applications.
  • Provides fine gas dispersion through a fritted glass diffuser.
  • Compatible with 20 L gas scrubber columns.
  • Promotes efficient gas-liquid contact for improved scrubbing performance.
  • Porous frit generates fine bubbles to increase mass transfer.
Catalog No. 50-273-3575 Supplier Chemglass Life Sciences Supplier No. CG183056
Fritted gas dispersion adapter designed to introduce and evenly disperse incoming gas into the liquid phase of a 20 L gas scrubber, improving gas-liquid contact for scrubbing applications.
  • Provides fine gas dispersion through a fritted glass diffuser.
  • Compatible with 20 L gas scrubber columns.
  • Promotes efficient gas-liquid contact for improved scrubbing performance.
  • Porous frit generates fine bubbles to increase mass transfer.