Learn More
Chemglass Life Sciences Gas Dispersion Tube, Coarse Frit
SDP
Catalog No. CG20303
Encompass_Preferred
Gas Dispersion Tube, Coarse Frit. Hollow fritted cylinder at the end of an 8mm O.D. tube. Cylinder is approximately 12mm O.D. x 20mm long. Overall length is approximately 10 inches.
Catalog No. CG20303 Supplier Chemglass Life Sciences Supplier No. CG20303
Gas Dispersion Tube, Coarse Frit. Hollow fritted cylinder at the end of an 8mm O.D. tube. Cylinder is approximately 12mm O.D. x 20mm long. Overall length is approximately 10 inches.