Learn More
Chemglass Life Sciences Gas Dispersion Tube, Medium Frit
SDP
Catalog No. CG20302
Encompass_Preferred
Gas Dispersion Tube, Medium Frit. Hollow fritted cylinder at the end of an 8mm O.D. tube. Cylinder is approximately 12mm O.D. x 20mm long. Overall length is approximately 10 inches.
Catalog No. CG20302 Supplier Chemglass Life Sciences Supplier No. CG20302
Gas Dispersion Tube, Medium Frit. Hollow fritted cylinder at the end of an 8mm O.D. tube. Cylinder is approximately 12mm O.D. x 20mm long. Overall length is approximately 10 inches.