Learn More
Chemglass Life Sciences Gas Dispersion Tube, Micro, Medium Frit
SDP
Catalog No. CG20702
Encompass_Preferred
Gas Dispersion Tube, Micro, Medium Frit. Micro size gas dispersion tube having a frit 7mm O.D. x 12mm. Stem is 6mm O.D. x 100mm long.
Catalog No. CG20702 Supplier Chemglass Life Sciences Supplier No. CG20702
Gas Dispersion Tube, Micro, Medium Frit. Micro size gas dispersion tube having a frit 7mm O.D. x 12mm. Stem is 6mm O.D. x 100mm long.