Learn More
Chemglass Life Sciences Gas Dispersion Tube, Micro, Coarse Frit
SDP
Catalog No. CG20703
Encompass_Preferred
Micro size gas dispersion tube having a frit 7mm O. D. x 12mm. Stem is 6mm O. D. x 100mm long.
Catalog No. CG20703 Supplier Chemglass Life Sciences Supplier No. CG20703
Micro size gas dispersion tube having a frit 7mm O. D. x 12mm. Stem is 6mm O. D. x 100mm long.