Learn More
Chemglass Life Sciences 50mL Gas Evolution Measurement Apparatus
SDP
Catalog No. 501531248
Encompass_Preferred
50mL Gas Evolution Measurement Apparatus. Apparatus provides a convenient and accurate method for the determination of the concentration or purity of active metals and metal hydrides. Metals and metal hydrides that can be analyzed in this manner include borane-methyl sulfide complexes, borane tetrahydrofuran, sodium and lithium as dispersions in mineral oil, lithium aluminum hydride. These active metals and metal hydrides react rapidly and quantiatively with suitable hydrolytic solvents to produce hydrogen gas. All complete units supplied with a buret, a CG-3502-04, -10 to 260 thermometer, a CG-1042-01 24/40 thermometer adapter and a leveling bulb with a 24/40 top outer and a lower detachable hose connection. The leveling bulb capacity varies with the capacity of the buret and has a hose connection of 8mm O.D. Flexible tubing is not supplied with the apparatus.
Catalog No. 50-153-1248 Supplier Chemglass Life Sciences Supplier No. CG181801
50mL Gas Evolution Measurement Apparatus. Apparatus provides a convenient and accurate method for the determination of the concentration or purity of active metals and metal hydrides. Metals and metal hydrides that can be analyzed in this manner include borane-methyl sulfide complexes, borane tetrahydrofuran, sodium and lithium as dispersions in mineral oil, lithium aluminum hydride. These active metals and metal hydrides react rapidly and quantiatively with suitable hydrolytic solvents to produce hydrogen gas. All complete units supplied with a buret, a CG-3502-04, -10 to 260 thermometer, a CG-1042-01 24/40 thermometer adapter and a leveling bulb with a 24/40 top outer and a lower detachable hose connection. The leveling bulb capacity varies with the capacity of the buret and has a hose connection of 8mm O.D. Flexible tubing is not supplied with the apparatus.