Learn More
Glass Expansion Gas Fitting with Ratchet Nebulizer Connector for Shimadzu 9000/9800
SDP
Catalog No. 501907301
Encompass_Preferred
Direct Connect (DC) Nebulizer Gas Fitting Type 41. Contains Ratchet Connector Gas Line 14-28 x 500 Assembly (70-803-1505) and ICP Gas Inlet Fitting. For argon gas connection between DC nebulizer and instrument argon gas supply fitting. Suitable for A41- DC Nebulizers. Compatible with all Shimadzu ICP-OES and ICP-MS.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7301 Supplier Glass Expansion Supplier No. 708031311
Direct Connect (DC) Nebulizer Gas Fitting Type 41. Contains Ratchet Connector Gas Line 14-28 x 500 Assembly (70-803-1505) and ICP Gas Inlet Fitting. For argon gas connection between DC nebulizer and instrument argon gas supply fitting. Suitable for A41- DC Nebulizers. Compatible with all Shimadzu ICP-OES and ICP-MS.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001