Learn More
Perkin Elmer US LLC Gas Fitting with Ratchet Nebulizer Connector
Catalog No. 502104349
Encompass_Preferred
This gas fitting is for use with all Direct Connection (DC) nebulizers. Direct Connection nebulizers have a UniFit sample connector which slides easily over the sample arm and an argon connector configured to connect directly to your ICP instrument.
Catalog No. 50-210-4349 Supplier Perkin Elmer US LLC Supplier No. N0811308
This gas fitting is for use with all Direct Connection (DC) nebulizers. Direct Connection nebulizers have a UniFit sample connector which slides easily over the sample arm and an argon connector configured to connect directly to your ICP instrument.