Learn More
Harrick Plasma PlasmaFlo Gas Flow Mixer 115V
SDP
Catalog No. NC0714406
Encompass_Preferred
The PDC-FMG PlasmaFlo may be used with any of our plasma cleaners. The PlasmaFlo provides more precise, quantitative control of the gas flowrate and monitoring of vacuum pressure, both of which are beneficial for process consistency. A second gas source can be added without having to manually swap the gas line. Dual gas feeds into flowmeters for gas mixing or independent control of two process gases; 49 mL/min maximum flowrate for atmospheric air at 0 psig (± 2% full scale accuracy); Vacuum gauge sensor connects to plasma cleaner door; Digital meter displays pressure range of 1 mTorr to 760 Torr; Size: 8.5″ H x 10″ W x 8″ D; Weight: 7 lb.
Catalog No. NC0714406 Supplier Harrick Plasma Supplier No. PDCFMG
The PDC-FMG PlasmaFlo may be used with any of our plasma cleaners. The PlasmaFlo provides more precise, quantitative control of the gas flowrate and monitoring of vacuum pressure, both of which are beneficial for process consistency. A second gas source can be added without having to manually swap the gas line. Dual gas feeds into flowmeters for gas mixing or independent control of two process gases; 49 mL/min maximum flowrate for atmospheric air at 0 psig (± 2% full scale accuracy); Vacuum gauge sensor connects to plasma cleaner door; Digital meter displays pressure range of 1 mTorr to 760 Torr; Size: 8.5″ H x 10″ W x 8″ D; Weight: 7 lb.