Learn More
Chemglass Life Sciences Gas Flowmeter, Low Flow Rate, O2, 30-300mL / Minute
SDP
Catalog No. 501532227
Encompass
Gas flowmeters for use with CHEMcell Rocker Bioreactor System, CLS-1200-series.Available for low and high flow rate based cell culture requirements.Inlet and outlet connections are made via 1/4" hose barb fittings.Gas must be dried, filtered and regulated to less than 15psi.Flowmeters also include a triangular base with leveling feet.Please contact us for information on flowmeters with other rates.
Catalog No. 50-153-2227 Supplier Chemglass Life Sciences Supplier No. CLS143910
Gas flowmeters for use with CHEMcell Rocker Bioreactor System, CLS-1200-series.Available for low and high flow rate based cell culture requirements.Inlet and outlet connections are made via 1/4" hose barb fittings.Gas must be dried, filtered and regulated to less than 15psi.Flowmeters also include a triangular base with leveling feet.Please contact us for information on flowmeters with other rates.