Learn More
Chemglass Life Sciences Gas Flowmeter, High Flow Rate, CO2, 0.6-6 Liters / Minute
SDP
Catalog No. 501532230
Encompass
Gas Flowmeter, High Flow Rate, CO2, 0.6-6 Liters / Minute. Gas flowmeters for use with CHEMcell Rocker Bioreactor System, CLS-1200-series. Inlet and outlet connections are made via 1/4" hose barb fittings. Gas must be dried, filtered and regulated to less than 15psi. Flowmeters also include a triangular base with leveling feet.
Catalog No. 50-153-2230 Supplier Chemglass Life Sciences Supplier No. CLS143921
Gas Flowmeter, High Flow Rate, CO2, 0.6-6 Liters / Minute. Gas flowmeters for use with CHEMcell Rocker Bioreactor System, CLS-1200-series. Inlet and outlet connections are made via 1/4" hose barb fittings. Gas must be dried, filtered and regulated to less than 15psi. Flowmeters also include a triangular base with leveling feet.