Learn More
Grainger SURE FLAME Direct Fired Heater, Dual Fuel, 1,500,000 BTU, 120 V, 1 ea
Catalog No. 502777337
Encompass_Preferred
This direct-fired dual fuel heater delivers an output of 850,000 to 1,500,000 BTU/hr for high-capacity heating requirements. Designed for durability and performance in demanding environments, the unit features an upgraded control board with moisture-resistant potting and integrated diagnostics for reliable operation in temperatures as low as -40°F.
  • Dual fuel capability allows for quick changeover between vapor propane and natural gas
  • Equipped with a 1 HP motor delivering 7,000 CFM and a 350°F temperature rise
  • Features a rugged, stackable frame for efficient storage
  • Includes a 15 ft. remote thermostat with Hi/Lo/Off control
  • Supports stable operation with relay voltage tolerance down to 50V
  • Integrated lifting loop and carry bars simplify transport
  • CSA approved for safety and reliability
Catalog No. 50-277-7337 Supplier Grainger Supplier No. 180L32
Add to Cart
Add to Cart
Request a Quote
This direct-fired dual fuel heater delivers an output of 850,000 to 1,500,000 BTU/hr for high-capacity heating requirements. Designed for durability and performance in demanding environments, the unit features an upgraded control board with moisture-resistant potting and integrated diagnostics for reliable operation in temperatures as low as -40°F.
  • Dual fuel capability allows for quick changeover between vapor propane and natural gas
  • Equipped with a 1 HP motor delivering 7,000 CFM and a 350°F temperature rise
  • Features a rugged, stackable frame for efficient storage
  • Includes a 15 ft. remote thermostat with Hi/Lo/Off control
  • Supports stable operation with relay voltage tolerance down to 50V
  • Integrated lifting loop and carry bars simplify transport
  • CSA approved for safety and reliability