Learn More
MSC Value Collection Pressure Washer, 2.9 GPM, Gas, Hot Water, 2300 PSI
Catalog No. 502839263
Encompass
This hot water, gas-powered pressure washer features a General Triplex Ceramic Plunger Pump and a 50-foot hose. It delivers a flow rate of 2.9 GPM and a pressure of 2300 PSI, powered by a Vanguard engine with a 205.0 cc displacement. Designed for high-performance cleaning applications, it includes a 4 Wheel Box Frame for enhanced mobility.
  • Features a General Triplex Ceramic Plunger Pump
  • Includes a 50-foot hose
  • Provides a flow rate of 2.9 GPM
  • Delivers 2300 PSI of pressure
  • Powered by a gas engine
  • Designed for hot water applications
  • Equipped with a Vanguard engine
  • Has a 205.0 cc engine displacement
  • Built with a 4 Wheel Box Frame
Catalog No. 50-283-9263 Supplier MSC Supplier No. 73688913
Add to Cart
Add to Cart
Request a Quote
This hot water, gas-powered pressure washer features a General Triplex Ceramic Plunger Pump and a 50-foot hose. It delivers a flow rate of 2.9 GPM and a pressure of 2300 PSI, powered by a Vanguard engine with a 205.0 cc displacement. Designed for high-performance cleaning applications, it includes a 4 Wheel Box Frame for enhanced mobility.
  • Features a General Triplex Ceramic Plunger Pump
  • Includes a 50-foot hose
  • Provides a flow rate of 2.9 GPM
  • Delivers 2300 PSI of pressure
  • Powered by a gas engine
  • Designed for hot water applications
  • Equipped with a Vanguard engine
  • Has a 205.0 cc engine displacement
  • Built with a 4 Wheel Box Frame