Learn More
Cleatech GAS-IN PORT-PRESSURE RELIEF
SDP
Catalog No. NC0384477
Encompass
Gas-in Port: Push-to-Connect fitting 1/4 in. OD tubing- Gas-out: Pressure releve /Bleeding valve 1/4 in.NPT
Catalog No. NC0384477 Supplier Cleatech Supplier No. A21GPINB
Gas-in Port: Push-to-Connect fitting 1/4 in. OD tubing- Gas-out: Pressure releve /Bleeding valve 1/4 in.NPT