Learn More
Ace Glass, Inc. GAS INLET TUBE GAS INLET TUBE
SDP
Catalog No. 50976725
Encompass_Preferred
Gas Inlet Tube, for use with Short Path Distillation Apparatus. Apparatus is designed for small quantities where holdup between flask and receiver must be minimized, but where it is also desired to measure vapor temperature with an accuracy compared to large setups.
Catalog No. 50-976-725 Supplier Ace Glass, Inc. Supplier No. 931606
Gas Inlet Tube, for use with Short Path Distillation Apparatus. Apparatus is designed for small quantities where holdup between flask and receiver must be minimized, but where it is also desired to measure vapor temperature with an accuracy compared to large setups.