Learn More
Bellco Glass, Inc. Gas Liquid Additon Assembly ,45mm For Vertical Sidearm Vessels
SDP
Catalog No. 501944714
Encompass_Preferred
Glass liquid addition assembly for use on 45mm side arms. Assembly simply screws on and seals with a silicone liner. MADE IN THE USA One glass addition tube type 1 a Borosilicate One compression cap One front & rear nylon ferrule One compression fitting
Catalog No. 50-194-4714 Supplier Bellco Glass, Inc. Supplier No. 196400045
Glass liquid addition assembly for use on 45mm side arms. Assembly simply screws on and seals with a silicone liner. MADE IN THE USA One glass addition tube type 1 a Borosilicate One compression cap One front & rear nylon ferrule One compression fitting