Learn More
Chemglass Life Sciences 4-Place Gas Manifold, Glass Stpk
SDP
Catalog No. CG181401
Encompass_Preferred
4-Place Gas Manifold, Glass Stpk. Four port manifold with 2mm bore glass stopcocks on the ports. A “T”-bore stopcock is located 100mm from the end of the manifold and 40mm from the first port. The distance between the other 3 ports is 50mm with the last port being 75mm from the opposite end of the manifold. The entire manifold is constructed of capillary tubing having an O.D. of 8mm and an I.D. of 2mm.
Catalog No. CG181401 Supplier Chemglass Life Sciences Supplier No. CG181401
4-Place Gas Manifold, Glass Stpk. Four port manifold with 2mm bore glass stopcocks on the ports. A “T”-bore stopcock is located 100mm from the end of the manifold and 40mm from the first port. The distance between the other 3 ports is 50mm with the last port being 75mm from the opposite end of the manifold. The entire manifold is constructed of capillary tubing having an O.D. of 8mm and an I.D. of 2mm.