Learn More
Chemglass Life Sciences Gas Mixing Module
SDP
Catalog No. 501941156
Encompass
The CHEMcell Gas mixing Module is used for accurate gas mixing of two gases operated through a touchscreen display with a maximum flow rate of up to 1.1 Liters per minute.Unit is equipped with two Alicat gas flow controllers while built in back pressure sensors allow for safe operation in a disposable bag or bioreactor.Gas totalizers allow you to track the total flow during the cell culture run.
Catalog No. 50-194-1156 Supplier Chemglass Life Sciences Supplier No. CLS1200GMM
Add to Cart
Add to Cart
Request a Quote
The CHEMcell Gas mixing Module is used for accurate gas mixing of two gases operated through a touchscreen display with a maximum flow rate of up to 1.1 Liters per minute.Unit is equipped with two Alicat gas flow controllers while built in back pressure sensors allow for safe operation in a disposable bag or bioreactor.Gas totalizers allow you to track the total flow during the cell culture run.