Learn More
Beckman Coulter Beckman Coulter gas-permeable sealing foil for microtiter plates, 10/pack (M2P-F-GPR48-10)
Catalog No. NC0891834
Encompass
Beckman Coulter M2P-F-GPR48-10 is a gas-permeable sealing foil designed to reduce evaporation and protect cultures in microtiter plates. The disposable adhesive film allows sufficient gas exchange while minimizing liquid loss and contamination, improving reproducibility in small-volume assays.
  • Reduces evaporation from wells.
  • Allows gas exchange to support aerobic cultures.
  • Adhesive film provides a secure seal for microtiter plates.
  • Sold in packs of 10 foils.
  • Disposable single-use design.
Catalog No. NC0891834 Supplier Beckman Coulter Supplier No. M2PFGPR4810
Beckman Coulter M2P-F-GPR48-10 is a gas-permeable sealing foil designed to reduce evaporation and protect cultures in microtiter plates. The disposable adhesive film allows sufficient gas exchange while minimizing liquid loss and contamination, improving reproducibility in small-volume assays.
  • Reduces evaporation from wells.
  • Allows gas exchange to support aerobic cultures.
  • Adhesive film provides a secure seal for microtiter plates.
  • Sold in packs of 10 foils.
  • Disposable single-use design.