Learn More
MSC Simpson Pressure Washer, 3200 PSI, 2.8 GPM, HONDA GX200 Engine, CAT PUMPS Triplex Pump
Catalog No. 502838630
Encompass
This powerful cold water mobile pressure washing system is designed for commercial applications. It is perfectly suited for contract cleaners, mobile detailers, municipalities, and resorts, and is DOT certified as an over-the-road pressure washer system.
  • Equipped with an engine featuring low oil shutdown.
  • Reliable triplex plunger pump with external unloader and bypass hose.
  • Includes a 50 ft. dual braided high-pressure hose.
  • Comes with 5 quick connect nozzle tips for various applications.
  • Features 2000 lb. single axle construction for durability.
  • Large 100 gallon water tank with easy-to-read levels.
  • Integrated LED lighting for visibility.
  • Premium 6-ply tires for easy maneuverability on rugged terrain.
Catalog No. 50-283-8630 Supplier MSC Supplier No. 39114509
Add to Cart
Add to Cart
Request a Quote
This powerful cold water mobile pressure washing system is designed for commercial applications. It is perfectly suited for contract cleaners, mobile detailers, municipalities, and resorts, and is DOT certified as an over-the-road pressure washer system.
  • Equipped with an engine featuring low oil shutdown.
  • Reliable triplex plunger pump with external unloader and bypass hose.
  • Includes a 50 ft. dual braided high-pressure hose.
  • Comes with 5 quick connect nozzle tips for various applications.
  • Features 2000 lb. single axle construction for durability.
  • Large 100 gallon water tank with easy-to-read levels.
  • Integrated LED lighting for visibility.
  • Premium 6-ply tires for easy maneuverability on rugged terrain.