Learn More
MSC SIMPSON Pressure Washer, 4200 PSI, 4.0 GPM, Cold Water, Gas
Catalog No. 502838219
Encompass
The SIMPSON PowerShot PS4240 is a cold water professional gas pressure washer designed for the rigorous demands of cleaning professionals. It is ideal for tasks such as deck cleaning, wood restoration, paint preparation, and graffiti removal. This unit features a powerful GX390 engine, industrial triplex pump, and a durable welded steel frame with premium pneumatic tires for easy maneuverability.
  • 4200 PSI pressure rating
  • 4.0 GPM flow rate
  • Powerful GX390 engine with low oil shutdown feature
  • Industrial triplex pump with enhanced performance
  • 50 ft. steel-braided monster hose, kink and abrasion resistant
  • 13 in. premium pneumatic tires for increased mobility
  • Durable welded steel frame with powder-coated finish
  • 5 quick connect nozzle tips for various tasks
  • Designed for rigorous professional cleaning applications
Catalog No. 50-283-8219 Supplier MSC Supplier No. 39114491
Add to Cart
Add to Cart
Request a Quote
The SIMPSON PowerShot PS4240 is a cold water professional gas pressure washer designed for the rigorous demands of cleaning professionals. It is ideal for tasks such as deck cleaning, wood restoration, paint preparation, and graffiti removal. This unit features a powerful GX390 engine, industrial triplex pump, and a durable welded steel frame with premium pneumatic tires for easy maneuverability.
  • 4200 PSI pressure rating
  • 4.0 GPM flow rate
  • Powerful GX390 engine with low oil shutdown feature
  • Industrial triplex pump with enhanced performance
  • 50 ft. steel-braided monster hose, kink and abrasion resistant
  • 13 in. premium pneumatic tires for increased mobility
  • Durable welded steel frame with powder-coated finish
  • 5 quick connect nozzle tips for various tasks
  • Designed for rigorous professional cleaning applications