Learn More
Cole-Parmer Drierite 27068 L68GP Gas Purifier, 300 L/hr, 100 psi
Supplier:  Cole-Parmer 0141850
Encompass_Preferred
Allows a maximum flow rate of 300 LPH and a maximum working pressure of 100 psi. Half of the polycarbonate column is filled with 250 mL of indicating Drierite producing a terminal dryness of 0.005 mg/liter. The other half contains 250 mL of 5 Å molecular sieves for removal of trace impurities. Total capacity is 500 mL. Both inlet and outlet fittings are 316 stainless steel, 1/8" Swagelock fittings. A 40-µm, 316 stainless steel outlet screen assembly prevents particulate migration. Drierite may be regenerated, or order the refill kit.
Catalog No. NC1632966