Learn More
AccuStandard, Inc GAS REFINE AROMATICS CAL STAND
SDP
Catalog No. NC1091971
Encompass
Gasoline Refinery Aromatics Calibration Set. 5 mL. Varied conc. in Isooctane.
Catalog No. NC1091971 Supplier AccuStandard, Inc Supplier No. D5769CALISSET
Gasoline Refinery Aromatics Calibration Set. 5 mL. Varied conc. in Isooctane.