Learn More
MATHESON TRI GAS Model 1250 Series Dual-Stage Brass Regulator
Catalog No. NC0606171
Encompass
High purity regulators constructed for use with non-corrosive gases. Stainless Steel Diaphragms. Delivery pressure range 4-50 psig. Delivery pressure gage 0-100 psig.
Catalog No. NC0606171 Supplier MATHESON TRI GAS Supplier No. 1252CGA
High purity regulators constructed for use with non-corrosive gases. Stainless Steel Diaphragms. Delivery pressure range 4-50 psig. Delivery pressure gage 0-100 psig.