Learn More
Grainger Dormont Gas Safety Kit, Stainless Steel, 36 in. Length, 1/2 in. Diameter, 1 ea.
Catalog No. 502778498
Encompass_Preferred
This gas safety kit is designed for use with moveable commercial kitchen equipment. It features a 36 in. (91.4 cm) long flexible hose with a 1/2 in. inside diameter and includes a one-handed quick-disconnect fitting with an integrated thermal shut-off that automatically halts gas flow if internal temperatures exceed 350°F (177°C).
  • Features antimicrobial PVC coating over woven corrugated 304 stainless steel tubing
  • Includes a quick-disconnect fitting with push-to-connect and thermal shut-off functionality
  • Equipped with 360° rotational end fittings for flexible movement
  • Designed for indoor use with commercial gas plumbing
  • Certified to ANSI Z21.69 and CSA 6.16 standards
  • Includes restraining cable and mounting hardware
Catalog No. 50-277-8498 Supplier Grainger Supplier No. 18D839
This gas safety kit is designed for use with moveable commercial kitchen equipment. It features a 36 in. (91.4 cm) long flexible hose with a 1/2 in. inside diameter and includes a one-handed quick-disconnect fitting with an integrated thermal shut-off that automatically halts gas flow if internal temperatures exceed 350°F (177°C).
  • Features antimicrobial PVC coating over woven corrugated 304 stainless steel tubing
  • Includes a quick-disconnect fitting with push-to-connect and thermal shut-off functionality
  • Equipped with 360° rotational end fittings for flexible movement
  • Designed for indoor use with commercial gas plumbing
  • Certified to ANSI Z21.69 and CSA 6.16 standards
  • Includes restraining cable and mounting hardware