Learn More
Grainger Dormont Gas Safety Kit, Stainless Steel, 1 in. Diameter, 36 in. Length, 1 ea
Catalog No. 502778432
Encompass_Preferred
This gas safety kit is designed for use with moveable commercial kitchen equipment. It features a flexible stainless steel hose assembly with a blue antimicrobial PVC coating that melts at 350°F (177°C) to provide a safety measure. The kit includes a one-handed quick-disconnect fitting with an integrated thermal shut-off design, ensuring gas flow is automatically terminated if internal temperatures exceed 350°F (177°C).
  • 36 in. (914 mm) length with 1 in. inside diameter
  • Constructed from 304 stainless steel tubing with multi-strand stainless steel wire braiding
  • Includes quick-disconnect fitting, elbow, and coiled restraining cable with mounting hardware
  • 360° rotational end fittings
  • 379,000 BTU/hr minimum flow capacity
  • Certified to ANSI Z21.69/CSA 6.16, ANSI Z21.41/CSA 6.9, and NSF/ANSI 169 standards
  • Indoor use only
  • Limited lifetime warranty
Catalog No. 50-277-8432 Supplier Grainger Supplier No. 18D841
This gas safety kit is designed for use with moveable commercial kitchen equipment. It features a flexible stainless steel hose assembly with a blue antimicrobial PVC coating that melts at 350°F (177°C) to provide a safety measure. The kit includes a one-handed quick-disconnect fitting with an integrated thermal shut-off design, ensuring gas flow is automatically terminated if internal temperatures exceed 350°F (177°C).
  • 36 in. (914 mm) length with 1 in. inside diameter
  • Constructed from 304 stainless steel tubing with multi-strand stainless steel wire braiding
  • Includes quick-disconnect fitting, elbow, and coiled restraining cable with mounting hardware
  • 360° rotational end fittings
  • 379,000 BTU/hr minimum flow capacity
  • Certified to ANSI Z21.69/CSA 6.16, ANSI Z21.41/CSA 6.9, and NSF/ANSI 169 standards
  • Indoor use only
  • Limited lifetime warranty