Learn More
Grainger Dormont Gas Connector Hose, 3/4 in., 36 in. Long, Stainless Steel, 1 ea.
Catalog No. 502778431
Encompass_Preferred
The 1675BPQR36 moveable gas connector is designed for use with commercial cooking appliances. This 36 in. coupled assembly features a 3/4 in. inner diameter 304 stainless steel corrugated tube for durability, protected by a tight-weave stainless steel braid and a blue antimicrobial PVC coating that inhibits the growth of bacteria, mold, and mildew. It is engineered for reliability in foodservice environments with a minimum flow capacity of 218,000 BTU/hr.
  • Features rotational technology fittings on both ends that rotate 360 degrees
  • Includes a push-to-connect quick-disconnect with a built-in thermal shutoff feature
  • Provides one-handed quick-disconnect functionality
  • Certified to ANSI Z21.69/CSA 6.16 and NSF/ANSI 169 standards
  • Operating temperature range of -40°F to +350°F
  • Includes a restraining device with mounting hardware
  • Backed by a limited lifetime warranty
Catalog No. 50-277-8431 Supplier Grainger Supplier No. 18D840
The 1675BPQR36 moveable gas connector is designed for use with commercial cooking appliances. This 36 in. coupled assembly features a 3/4 in. inner diameter 304 stainless steel corrugated tube for durability, protected by a tight-weave stainless steel braid and a blue antimicrobial PVC coating that inhibits the growth of bacteria, mold, and mildew. It is engineered for reliability in foodservice environments with a minimum flow capacity of 218,000 BTU/hr.
  • Features rotational technology fittings on both ends that rotate 360 degrees
  • Includes a push-to-connect quick-disconnect with a built-in thermal shutoff feature
  • Provides one-handed quick-disconnect functionality
  • Certified to ANSI Z21.69/CSA 6.16 and NSF/ANSI 169 standards
  • Operating temperature range of -40°F to +350°F
  • Includes a restraining device with mounting hardware
  • Backed by a limited lifetime warranty