Learn More
Accuflow Systems Saint-Gobain Gas Sampling Bag, FEP, 6 x 10 in
Catalog No. NC2327747
Encompass_Preferred
Saint-Gobain Chemware FEP gas sampling bag for collection, transport, and short-term storage of gaseous samples. Constructed from chemically inert FEP film and heat-sealed on all sides to minimize permeability and maintain sample integrity. Typical applications include air quality monitoring, stack gas sampling, and calibration gas collection.
  • Made from 5 mil FEP fluoropolymer film.
  • Dimensions 6 x 10 in, capacity 1.2 L.
  • Valve options include nickel-plated brass on/off fitting or PTFE/silicone septum.
  • Heat-sealed on all four sides for leak-resistant containment.
  • Low permeability to limit sample loss during storage and transport.
  • Individually packaged; suitable for corrosive or reactive sample matrices.
Catalog No. NC2327747 Supplier Accuflow Systems Supplier No. D1075001
Saint-Gobain Chemware FEP gas sampling bag for collection, transport, and short-term storage of gaseous samples. Constructed from chemically inert FEP film and heat-sealed on all sides to minimize permeability and maintain sample integrity. Typical applications include air quality monitoring, stack gas sampling, and calibration gas collection.
  • Made from 5 mil FEP fluoropolymer film.
  • Dimensions 6 x 10 in, capacity 1.2 L.
  • Valve options include nickel-plated brass on/off fitting or PTFE/silicone septum.
  • Heat-sealed on all four sides for leak-resistant containment.
  • Low permeability to limit sample loss during storage and transport.
  • Individually packaged; suitable for corrosive or reactive sample matrices.