Learn More
Chemglass Life Sciences 3L Gas Scrubber, Complete (Support Stand Sold Separately)
SDP
Catalog No. 50872430
Encompass_Preferred
3L Gas Scrubber, Complete (Support Stand Sold Separately). Laboratory scale gas scrubber for use in many air and exhaust cleanup tasks. The 3 liter is a compact design and will fit into any laboratory hood, or can be used in walk in hoods when assembled on our CG-1951-100 mobile or CG-1950-100 bench-top reactor stands.Gas scrubbing takes place when the exhaust enters the inlet in the round bottom flask and travels upward through the absorption column. The scrubbing fluid is pumped from the round bottom flask upwards into the shower head adapter and then flows counter-current to the exhaust gas through the packed column thereby causing intensive contact between the two. Scrubber can be used by creating either a positive flow through the reaction vessel using a carrier such as nitrogen, or by connection of a vacuum source to the column adapter.
Catalog No. 50-872-430 Supplier Chemglass Life Sciences Supplier No. CG183010
Add to Cart
Add to Cart
Request a Quote
3L Gas Scrubber, Complete (Support Stand Sold Separately). Laboratory scale gas scrubber for use in many air and exhaust cleanup tasks. The 3 liter is a compact design and will fit into any laboratory hood, or can be used in walk in hoods when assembled on our CG-1951-100 mobile or CG-1950-100 bench-top reactor stands.Gas scrubbing takes place when the exhaust enters the inlet in the round bottom flask and travels upward through the absorption column. The scrubbing fluid is pumped from the round bottom flask upwards into the shower head adapter and then flows counter-current to the exhaust gas through the packed column thereby causing intensive contact between the two. Scrubber can be used by creating either a positive flow through the reaction vessel using a carrier such as nitrogen, or by connection of a vacuum source to the column adapter.