Learn More
Chemglass Life Sciences Support Stand for 20L Gas Scrubber, Walk in Hood, Mobile
SDP
Catalog No. 501311105
Encompass_Preferred
Mobile support stand for 20L gas scrubbers for use in walk in hoods only.Requires:(2) CG-9205-06(2) CG-9253-10for CG-1830-50 Gas Scrubber System
Catalog No. 50-131-1105 Supplier Chemglass Life Sciences Supplier No. CG183060
Mobile support stand for 20L gas scrubbers for use in walk in hoods only.Requires:(2) CG-9205-06(2) CG-9253-10for CG-1830-50 Gas Scrubber System