Learn More
Grainger ASRaymond Gas Spring, M6 Thread, 186 lbf Force, 7 63/64 in. Length, Black Oxide, 2/pk
Catalog No. 502773808
Encompass_Preferred
This item has to be ordered in multiples of 2 per supplier requirements.
This standard gas spring is designed for reliable motion control applications, featuring a M6 thread size and a force rating of 186 lbf. Engineered for durability and performance, this component provides consistent load-carrying characteristics throughout its operational life.
  • Extended length of 7 63/64 in.
  • Stroke length of 3 1/8 in.
  • Black oxide finish for corrosion resistance
  • Operating temperature range from 32 °F to 176 °F
  • Mounting orientation supports drop in, threaded bottom, and top configurations
  • Non-locking design with adjustable resistance
Catalog No. 50-277-3808 Supplier Grainger Supplier No. 18AY62
This item has to be ordered in multiples of 2 per supplier requirements.
This standard gas spring is designed for reliable motion control applications, featuring a M6 thread size and a force rating of 186 lbf. Engineered for durability and performance, this component provides consistent load-carrying characteristics throughout its operational life.
  • Extended length of 7 63/64 in.
  • Stroke length of 3 1/8 in.
  • Black oxide finish for corrosion resistance
  • Operating temperature range from 32 °F to 176 °F
  • Mounting orientation supports drop in, threaded bottom, and top configurations
  • Non-locking design with adjustable resistance