Learn More
Grainger ASRaymond Gas Spring, Black Oxide, 2 5/8 in., 74 lbf., M6
Catalog No. 502773964
Encompass_Preferred
This standard gas spring is designed for controlled lifting and dampening applications. Featuring a durable black oxide finish, it provides consistent performance in a variety of industrial settings with an adjustable resistance capacity.
  • Force rating of 74 lbf.
  • Extended length of 2 5/8 in. and compressed length of 2 1/8 in.
  • M6 thread size for versatile mounting.
  • Operating temperature range of 32°F to 176°F.
  • Stroke length of 1/2 in.
  • Cylinder diameter of 0.47 in.
  • Medium duty design.
Catalog No. 50-277-3964 Supplier Grainger Supplier No. 18AY23
This standard gas spring is designed for controlled lifting and dampening applications. Featuring a durable black oxide finish, it provides consistent performance in a variety of industrial settings with an adjustable resistance capacity.
  • Force rating of 74 lbf.
  • Extended length of 2 5/8 in. and compressed length of 2 1/8 in.
  • M6 thread size for versatile mounting.
  • Operating temperature range of 32°F to 176°F.
  • Stroke length of 1/2 in.
  • Cylinder diameter of 0.47 in.
  • Medium duty design.