Learn More
Vici Precision Sampling, Inc. GAS SYRING 2ML 1EA
Catalog No. NC9704060
Encompass_Preferred
Gas Syringe A-2 2ml RN .029 in. x .012 in. x 2 in. bevel, open end
Catalog No. NC9704060 Supplier Vici Precision Sampling, Inc. Supplier No. 050034
Gas Syringe A-2 2ml RN .029 in. x .012 in. x 2 in. bevel, open end