Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DSI LLC Gas Syringe Magnum 500ml RN .035" x .023"x 2" bevel open end

Catalog No. 501871621
Encompass_Preferred

Gas Syringe Magnum 500ml RN .035" x .023"x 2" bevel open end

Catalog No. 50-187-1621 Supplier DSI LLC Supplier No. DS060042
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gas Syringe Magnum 500ml RN .035" x .023"x 2" bevel open end

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.