Learn More
Binder Inc Gas tank changer
SDP
Supplier:  Binder Inc 80122344
Encompass_Preferred
Gas tank changer - external, BINDER Gas Supply Service, for connecting 2 gas tanks (CO2, N2 or O2), with audible and visual alarms, as well as zero-voltage alarm output
Catalog No. 80-122-344