Learn More
BURKLE INC Gas-tight barrel pump PP, connection 2x3/4"
Catalog No. 5000025548
Encompass_Preferred
During transfer with normal pumps, it is impossible to prevent vapours escaping into the atmosphere. The gas-shuttle method avoids this. Any vapours produced are returned from the container which is to be filled to the original container via the gas-shuttle pipe. The gas pipe and the liquid pipe are additionally provided with stopcocks.
Gas-tight pumps are used for substances that have a strong smell such as ammonia.
Catalog No. 50-000-25548 Supplier BURKLE INC Supplier No. 56101040
During transfer with normal pumps, it is impossible to prevent vapours escaping into the atmosphere. The gas-shuttle method avoids this. Any vapours produced are returned from the container which is to be filled to the original container via the gas-shuttle pipe. The gas pipe and the liquid pipe are additionally provided with stopcocks.
Gas-tight pumps are used for substances that have a strong smell such as ammonia.