Learn More
BINDER™ Gas-tight Option Model
SDP
Catalog No. 08111716 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Steel
Catalog No. Material
08-111-716 Steel
1 options
Catalog No. 08-111-716 Supplier BINDER™ Supplier No. 80121045
Add to Cart
Add to Cart
For Use With: BF 115
  • Includes additional measures for the greatest possible impermeability to gas
  • Not in conjunction with access port or interior lighting option
Specifications
Description Gas-tight option model - Incl. additional measures for the greatest possible impermeability to gas; not in conjunction with access port or interior lighting option
For Use With (Equipment) BINDER Chambers BF 115
Material Steel
Type Gas-tight Option Model